3-Chloro-5-(ethylcarbamoyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-5-(ethylcarbamoyl)phenyl)boronic acid

A248077-100MG A248077-1G A248077-250MG
76874-556EA 37.14 USD
76874-556 76874-558 76874-560
3-Chloro-5-(ethylcarbamoyl)phenylboronic acid 96%
3-Chloro-5-(ethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClNO₃
MW: 227.4500 g/mol
MDL Number: MFCD09800857
CAS Number: 957120-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR