(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-methoxy-5-methylphenylboronic acid

A247994-1G A247994-250MG A247994-5G
76874-550EA 218 USD
76874-550 76874-552 76874-554
(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid 98%
(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD18383901
CAS Number: 1207428-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR