Methyl 4-(aminomethyl)-3-chlorobenzoate 95%

Supplier: Ambeed
Warning

A247142-100MG A247142-1G A247142-250MG
76874-532EA 23.34 USD
76874-532 76874-534 76874-536
Methyl 4-(aminomethyl)-3-chlorobenzoate 95%
Methyl 4-(aminomethyl)-3-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD10566566
CAS Number: 940062-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR