3-Amino-5-bromo-2-nitropyridine 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-nitropyridin-3-amine

A246693-100MG A246693-250MG A246693-5G A246693-1G
76874-522EA 22 USD
76874-522 76874-524 76874-526 77612-384
3-Amino-5-bromo-2-nitropyridine 96%
3-Amino-5-bromo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrN₃O₂
MW: 218.008 g/mol
MDL Number: MFCD11040269
CAS Number: 433226-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR