Oxetan-3-ylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Oxetan-3-yl)acetic acid

A246503-250MG A246503-1G A246503-100MG
76874-516EA 165.51 USD
76874-516 77568-396 77672-200
Oxetan-3-ylacetic acid 98%
Oxetan-3-ylacetic acid

Precaución: For research use only.

Formula: C₅H₈O₃
MW: 116.115 g/mol
MDL Number: MFCD19442256
CAS Number: 1310381-54-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR