3-Chloro-5-hydroxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A246459-100MG A246459-1G A246459-250MG
76874-510EA 37.14 USD
76874-510 76874-512 76874-514
3-Chloro-5-hydroxyphenylboronic acid pinacol ester 98%
3-Chloro-5-hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BClO₃
MW: 254.5200 g/mol
MDL Number: MFCD12546581
CAS Number: 960388-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR