2,5-Dichloro-4-methoxypyridine 95%

Supplier: Ambeed
Warning

A245059-100MG A245059-1G A245059-250MG
76874-502EA 29.71 USD
76874-502 76874-504 76874-506
2,5-Dichloro-4-methoxypyridine 95%
2,5-Dichloro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
MDL Number: MFCD13185541
CAS Number: 1122090-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR