1-Boc-indole-2-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A244499-1G A244499-5G A244499-100MG A244499-250MG A244499-25G
76874-498EA 41 USD
76874-498 76874-500 77883-650 77883-651 77883-652
1-Boc-indole-2-boronic acid pinacol ester 95%
1-Boc-indole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₆BNO₄
MW: 343.2251 g/mol
MDL Number: MFCD09838985
CAS Number: 1072944-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR