Ethyl 2-(6-((tert-butoxycarbonyl)amino)pyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A244406-100MG A244406-1G A244406-250MG A244406-5G
76874-490EA 40 USD
76874-490 76874-492 76874-494 76874-496
Ethyl 2-(6-((tert-butoxycarbonyl)amino)pyridin-2-yl)acetate 95%
Ethyl 2-(6-((tert-butoxycarbonyl)amino)pyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₄
MDL Number: MFCD10698587
CAS Number: 408365-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR