2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone 97%

Supplier: Ambeed
Warning

A243745-5G A243745-1G
76874-470EA 366.02 USD
76874-470 77568-368
2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone 97%
2,2,2-Trifluoro-1-(piperazin-1-yl)ethanone

Precaución: For research use only.

Formula: C₆H₉F₃N₂O
MW: 182.14 g/mol
MDL Number: MFCD07782176
CAS Number: 6511-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR