5-Bromo-4-fluoro-2-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A243592-250MG A243592-1G
76874-466EA 11 USD
76874-466 77672-176
5-Bromo-4-fluoro-2-nitrobenzaldehyde 95%
5-Bromo-4-fluoro-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₃
MDL Number: MFCD00456499
CAS Number: 213382-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR