(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-Bis(alpha-methylbenzyl)amine Hydrochloride, (R,R)-(+)-Bis(1-phenylethyl)amine Hydrochloride

A242137-10G A242137-250MG A242137-25G A242137-5G A242137-1G
76874-434EA 81 USD
76874-434 76874-436 76874-438 76874-440 77672-168
(R,R)-(+)-Bis(ɑ-methylbenzyl)amine hydrochloride 98%
(R,R)-(+)-Bis(alpha-methylbenzyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₉N
MDL Number: MFCD00216672
CAS Number: 82398-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR