Benzo[d][1,2,3]thiadiazol-6-amine 97%

Supplier: Ambeed
Warning

A241761-100MG A241761-250MG
76874-428EA 144.29 USD
76874-428 76874-430
Benzo[d][1,2,3]thiadiazol-6-amine 97%
Benzo[d][1,2,3]thiadiazol-6-amine

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MW: 151.18 g/mol
MDL Number: MFCD20691248
CAS Number: 41972-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR