4-(4-Bromo-1,3-thiazol-2-yl)morpholine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Bromothiazol-2-yl)morpholine, 4-Bromo-2-(morpholin-4-yl)-1,3-thiazole

A240756-250MG A240756-1G
76874-408EA 24.41 USD
76874-408 77550-550
4-(4-Bromo-1,3-thiazol-2-yl)morpholine 98%
4-(4-Bromo-1,3-thiazol-2-yl)morpholine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂OS
MW: 249.128 g/mol
MDL Number: MFCD09607696
CAS Number: 1017781-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR