Dapsone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
A240646-100G
A240646-1MG
A240646-25G
A240646-500G
A240646-1000G
76874-400EA
9
USD
76874-400
76874-402
76874-404
76874-406
77593-536
Dapsone 98%
Dapsone
Precaución: for Research Use Only
Formula:
C₁₂H₁₂N₂O₂S MW: 248.31 g/mol |
MDL Number:
MFCD00007887 CAS Number: 80-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...