N-(5-Bromopyrimidin-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A239140-100MG A239140-10G A239140-1G A239140-250MG A239140-5G
76874-374EA 26.53 USD
76874-374 76874-376 76874-378 76874-380 76874-382
N-(5-Bromopyrimidin-2-yl)acetamide 98%
N-(5-Bromopyrimidin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O
MW: 216.03 g/mol
MDL Number: MFCD12546524
CAS Number: 180530-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR