3-(Methylthio)-6,7-dihydrobenzo[c]thiophen-4(5H)-one 95%

Supplier: Ambeed
Warning

A238890-100MG A238890-1G A238890-250MG
76874-368EA 94.43 USD
76874-368 76874-370 76874-372
3-(Methylthio)-6,7-dihydrobenzo[c]thiophen-4(5H)-one 95%
3-(Methylthio)-6,7-dihydrobenzo[c]thiophen-4(5H)-one

Precaución: for Research Use Only

Formula: C₉H₁₀OS₂
MW: 198.30 g/mol
MDL Number: MFCD00107351
CAS Number: 168279-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR