2-Bromo-4-chloro-6-methoxyaniline 98%

Supplier: Ambeed
Warning

A238767-100MG A238767-1G A238767-250MG A238767-5G
76874-360EA 10 USD
76874-360 76874-362 76874-364 76874-366
2-Bromo-4-chloro-6-methoxyaniline 98%
2-Bromo-4-chloro-6-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MDL Number: MFCD18311825
CAS Number: 1261895-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR