4-Chloro-o-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-5-chlorotoluene, 4-Chloro-2-methylaniline, 4-Chloro-o-toluidine, Fast Red TR Base

A238447-25G A238447-5G
76874-356EA 12 USD
76874-356 76874-358
4-Chloro-o-toluidine 98%
4-Chloro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MDL Number: MFCD00007842
CAS Number: 95-69-2
UN: 2239
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR