5-Bromo-2-methylamino-3-nitro-4-picoline 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-methyl-2-methylamino-3-nitropyridine, 5-Bromo-N,4-dimethyl-3-nitropyridin-2-amine

A237518-100MG A237518-1G
76874-350EA 5 USD
76874-350 76874-352
5-Bromo-2-methylamino-3-nitro-4-picoline 95%
5-Bromo-2-methylamino-3-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₇H₈BrN₃O₂
MW: 246.1 g/mol
MDL Number: MFCD08692210
CAS Number: 155790-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR