Dichlorobis(2-ethylhexyl)silane 95%

Supplier: Ambeed
Danger

A233672-1G A233672-25G A233672-5G
76874-304EA 26 USD
76874-304 76874-306 76874-308
Dichlorobis(2-ethylhexyl)silane 95%
Dichlorobis(2-ethylhexyl)silane

Precaución: for Research Use Only

Formula: C₁₆H₃₄Cl₂Si
MDL Number: MFCD22370170
CAS Number: 1089687-03-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR