2,6-Bis(benzyloxy)-3-bromopyridine 96%
Supplier: Ambeed
|
Warning
|
A233337-100G
A233337-100MG
A233337-10G
A233337-1G
A233337-250MG
A233337-25G
A233337-5G
76874-288EA
180
USD
76874-288
76874-290
76874-292
76874-294
76874-296
76874-298
76874-300
2,6-Bis(benzyloxy)-3-bromopyridine 96%
2,6-Bis(benzyloxy)-3-bromopyridine
Precaución: for Research Use Only
Formula:
C₁₉H₁₆BrNO₂ |
MDL Number:
MFCD18434484 CAS Number: 16727-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...