5,7-Dimethoxychroman-4-one 97%

Supplier: Ambeed
Warning

A232310-100MG A232310-1G
76874-274EA 53.05 USD
76874-274 76874-276
5,7-Dimethoxychroman-4-one 97%
5,7-Dimethoxychroman-4-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD12079198
CAS Number: 54107-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR