2-(3-Bromo-4-fluorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-fluorophenylacetonitrile

A232105-100G A232105-10G A232105-1G A232105-25G
76874-266EA 688.53 USD
76874-266 76874-268 76874-270 76874-272
2-(3-Bromo-4-fluorophenyl)acetonitrile 97%
2-(3-Bromo-4-fluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.037 g/mol
MDL Number: MFCD08458154
CAS Number: 501420-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR