1,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A229241-100MG A229241-1G A229241-250MG A229241-25G A229241-5G
76874-224EA 10.61 USD
76874-224 77672-034 77672-036 77672-038 77672-040
1,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%
1,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
MW: 222.09 g/mol
MDL Number: MFCD12400853
CAS Number: 1047644-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR