Methyl 5-bromo-2,4-dimethoxybenzoate 95%

Supplier: Ambeed
Warning

A228976-1G A228976-25G A228976-5G A228976-10G
76874-214EA 13 USD
76874-214 76874-216 76874-218 77672-028
Methyl 5-bromo-2,4-dimethoxybenzoate 95%
Methyl 5-bromo-2,4-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₄
MDL Number: MFCD05154996
CAS Number: 39503-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR