Methyl 2-((4-hydroxyphenyl)amino)acetate hydrochloride 95%

Supplier: Ambeed
Warning

A228191-100MG A228191-1G A228191-250MG
76874-184EA 10 USD
76874-184 76874-186 76874-188
Methyl 2-((4-hydroxyphenyl)amino)acetate hydrochloride 95%
Methyl 2-((4-hydroxyphenyl)amino)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₃
MDL Number: MFCD22421599
CAS Number: 113210-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR