2,2-Dimethylcyclohexane-1,3-dione 98%

Supplier: Ambeed
Warning

A228118-250MG A228118-25G A228118-5G A228118-1G
76874-178EA 29.71 USD
76874-178 76874-180 76874-182 77672-018
2,2-Dimethylcyclohexane-1,3-dione 98%
2,2-Dimethylcyclohexane-1,3-dione

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD14708261
CAS Number: 562-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR