tert-Butyl 2-(1-methylhydrazinyl)acetate 97%

Supplier: Ambeed
Warning

A227549-250MG A227549-5G A227549-100MG A227549-1G
76874-176EA 71.09 USD
76874-176 77568-210 77672-008 77672-010
tert-Butyl 2-(1-methylhydrazinyl)acetate 97%
tert-Butyl 2-(1-methylhydrazinyl)acetate

Precaución: For research use only.

Formula: C₇H₁₆N₂O₂
MW: 160.21 g/mol
MDL Number: MFCD20266159
CAS Number: 144036-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR