(3-Hydroxynaphthalen-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A226592-100MG A226592-1G A226592-250MG A226592-25G
76874-146EA 14 USD
76874-146 76874-148 76874-150 77885-564
(3-Hydroxynaphthalen-2-yl)boronic acid 98%
(3-Hydroxynaphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BO₃
MDL Number: MFCD18417836
CAS Number: 849404-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR