5-Bromobenzo[d]isothiazole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A226367-100MG A226367-1G A226367-250MG
76874-124EA 127.31 USD
76874-124 76874-126 76874-128
5-Bromobenzo[d]isothiazole-3-carboxylic acid 97%
5-Bromobenzo[d]isothiazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂S
MW: 258.09 g/mol
MDL Number: MFCD08706004
CAS Number: 677304-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR