4-Bromo-3-fluoro-2-nitroaniline 97%

Supplier: Ambeed
Warning

A225631-1G A225631-250MG A225631-5G A225631-25G
76874-094EA 20 USD
76874-094 76874-096 76874-098 77593-464
4-Bromo-3-fluoro-2-nitroaniline 97%
4-Bromo-3-fluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O₂
MDL Number: MFCD07368682
CAS Number: 886762-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR