3-Isopropylbenzaldehyde 98%

Supplier: Ambeed
Warning

A225491-100MG A225491-1G A225491-5G
76874-088EA 12.74 USD
76874-088 76874-090 76874-092
3-Isopropylbenzaldehyde 98%
3-Isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.20 g/mol
MDL Number: MFCD08062435
CAS Number: 34246-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR