5-Acetyl-2-fluorobenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 5-Acetyl-2-fluorophenylboronic acid pinacol ester

A225316-1G A225316-250MG A225316-25G A225316-5G
76874-080EA 40.32 USD
76874-080 76874-082 76874-084 76874-086
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester 98%
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₃
MW: 264.103 g/mol
MDL Number: MFCD16996292
CAS Number: 765916-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR