4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Ambeed
Danger

Synonyms: 2-(4-(Bromomethyl)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A223927-1G A223927-250MG A223927-5G
76874-050EA 166.57 USD
76874-050 76874-052 76874-054
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 96%
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.989 g/mol
MDL Number: MFCD18434471
CAS Number: 1029439-49-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR