(1R,4R)-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate 95%

Supplier: Ambeed
Warning

A223324-1G A223324-250MG A223324-5G A223324-100MG
76874-026EA 144.29 USD
76874-026 76874-028 76874-030 77593-454
(1R,4R)-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate 95%
(1R,4R)-tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.262 g/mol
MDL Number: MFCD01321293
CAS Number: 134003-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR