3-Amino-2-bromo-5-chloropyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-chloro-3-pyridylamine
A222813-10G
A222813-1G
A222813-25G
76874-018EA
37.14
USD
76874-018
76874-020
76874-022
3-Amino-2-bromo-5-chloropyridine 98%
3-Amino-2-bromo-5-chloropyridine
Precaución: for Research Use Only
Formula:
C₅H₄BrClN₂ MW: 207.46 g/mol |
MDL Number:
MFCD07368976 CAS Number: 90902-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...