2-(Chloromethyl)-3-fluoropyridine 95%

Supplier: Ambeed
Danger

A222328-100MG A222328-1G A222328-250MG
76873-994EA 47.75 USD
76873-994 76873-996 76873-998
2-(Chloromethyl)-3-fluoropyridine 95%
2-(Chloromethyl)-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.562 g/mol
MDL Number: MFCD11520697
CAS Number: 149489-32-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR