4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol 97%

Supplier: Ambeed
Warning

A222015-100G A222015-25G A222015-5G
76873-988EA 104 USD
76873-988 76873-990 76873-992
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol 97%
4-Methyl-2,3,5,6-tetrafluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MDL Number: MFCD03093260
CAS Number: 79538-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR