Ethyl 3-hydroxy-2,2-dimethylpropanoate 97%
Supplier: Ambeed
|
Warning
|
A221949-100G
A221949-10G
A221949-1G
A221949-250MG
A221949-25G
76873-978EA
811.59
USD
76873-978
76873-980
76873-982
76873-984
76873-986
Ethyl 3-hydroxy-2,2-dimethylpropanoate 97%
Ethyl 3-hydroxy-2,2-dimethylpropanoate
Precaución: for Research Use Only
Formula:
C₇H₁₄O₃ MW: 146.184 g/mol |
MDL Number:
MFCD21643853 CAS Number: 14002-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...