5-(Bromomethyl)undecane 95%

Supplier: Ambeed
Warning

A221294-1G A221294-250MG A221294-25G A221294-5G
76873-964EA 15.92 USD
76873-964 76873-966 76873-968 77593-434
5-(Bromomethyl)undecane 95%
5-(Bromomethyl)undecane

Precaución: for Research Use Only

Formula: C₁₂H₂₅Br
MW: 249.24 g/mol
MDL Number: MFCD23115881
CAS Number: 85531-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR