2-(Bis-boc-amino)-5-pyrimidineboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (tert-butoxycarbonyl)(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)carbamate

A221252-1G A221252-5G
76873-960EA 64.72 USD
76873-960 76873-962
2-(Bis-boc-amino)-5-pyrimidineboronic acid pinacol ester 95%
2-(Bis-Boc-amino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₃₂BN₃O₆
MW: 421.2956 g/mol
MDL Number: MFCD12913985
CAS Number: 1190423-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR