3,4-Dichloro-2-methylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3,4-Dichloro-2-methylphenyl)boronic acid

A220752-100MG A220752-250MG
76873-946EA 95.49 USD
76873-946 76873-948
3,4-Dichloro-2-methylphenylboronic acid 95%
3,4-Dichloro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₂
MW: 204.85 g/mol
MDL Number: MFCD09475873
CAS Number: 957035-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR