Diethyl ethoxymethylenemalonate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-(ethoxymethylene)malonate

A220560-1000G A220560-100G A220560-25G A220560-500G A220560-5G
76873-936EA 46 USD
76873-936 76873-938 76873-940 76873-942 76873-944
Diethyl ethoxymethylenemalonate 98%
Diethyl ethoxymethylenemalonate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₅
MDL Number: MFCD00009148
CAS Number: 87-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR