tert-Butyl 7-bromo-3,4-dihydroquinoline-1(2H)-carboxylate 97%

Supplier: Ambeed
Warning

A219947-1G A219947-250MG A219947-5G A219947-100MG
76873-926EA 238.71 USD
76873-926 76873-928 77593-430 77671-960
tert-Butyl 7-bromo-3,4-dihydroquinoline-1(2H)-carboxylate 97%
tert-Butyl 7-bromo-3,4-dihydroquinoline-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₂
MW: 312.20 g/mol
MDL Number: MFCD12913865
CAS Number: 1187932-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR