N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-trans-4-hydroxy-L-proline Methyl Ester, (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-Butyl 2-Methyl Ester, 1-tert-Butyl 2-Methyl (2S,4R)-4-Hydroxypyrrolidine-1,2-dicarboxylate, Boc-Hyp-OMe, 1-(tert-Butyl) 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
A219851-1000G
A219851-100G
A219851-10G
A219851-25G
A219851-500G
A219851-5G
76873-914EA
155
USD
76873-914
76873-916
76873-918
76873-920
76873-922
76873-924
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₅ |
MDL Number:
MFCD00076981 CAS Number: 74844-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...