5-Bromo-2,4-dimethylaniline 97%

Supplier: Ambeed
Warning

A219532-1G A219532-250MG A219532-5G
76873-898EA 34 USD
76873-898 76873-900 76873-902
5-Bromo-2,4-dimethylaniline 97%
5-Bromo-2,4-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MDL Number: MFCD09038481
CAS Number: 69383-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR