4-Chloro-6-fluoronicotinaldehyde 97%

Supplier: Ambeed
Warning

A218304-100MG A218304-1G A218304-250MG
76873-862EA 22 USD
76873-862 77883-552 77883-553
4-Chloro-6-fluoronicotinaldehyde 97%
4-Chloro-6-fluoronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MDL Number: MFCD18257916
CAS Number: 1256824-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR