3-Amino-4-bromo-5-methylisoxazole 98%

Supplier: Ambeed
Warning

A218142-1G A218142-250MG A218142-5G
76873-860EA 24 USD
76873-860 77883-550 77883-551
3-Amino-4-bromo-5-methylisoxazole 98%
3-Amino-4-bromo-5-methylisoxazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂O
MDL Number: MFCD00052553
CAS Number: 5819-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR