2-Amino-4-bromo-6-nitrophenol 98%

Supplier: Ambeed
Danger

A217306-100MG A217306-250MG
76873-848EA 38 USD
76873-848 76873-850
2-Amino-4-bromo-6-nitrophenol 98%
2-Amino-4-bromo-6-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MDL Number: MFCD22206403
CAS Number: 139138-08-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR